Learn More
AdipoGen W-5 . hydrochloride (25 mg)
SDP
Catalog No. 502053749
Encompass_Preferred
Chemical. CAS: 61714-25-8. Formula: C16H22N2O2S . HCl. MW: 306.4 . 36.5. Calmodulin antagonist. Cell proliferation inhibitor. Ca2+/calmodulin-regulated phosphodiesterase (PDE I) inhibitor. Myosin light chain kinase (MLCK) inhibitor.
Catalog No. 50-205-3749 Supplier AdipoGen Supplier No. AGCR10003M025
Chemical. CAS: 61714-25-8. Formula: C16H22N2O2S . HCl. MW: 306.4 . 36.5. Calmodulin antagonist. Cell proliferation inhibitor. Ca2+/calmodulin-regulated phosphodiesterase (PDE I) inhibitor. Myosin light chain kinase (MLCK) inhibitor.