Learn More
AdipoGen W-5 . hydrochloride (5 mg)
SDP
Supplier:  AdipoGen AGCR10003M005
Encompass_Preferred
Chemical. CAS: 61714-25-8. Formula: C16H22N2O2S . HCl. MW: 306.4 . 36.5. Calmodulin antagonist. Cell proliferation inhibitor. Ca2+/calmodulin-regulated phosphodiesterase (PDE I) inhibitor. Myosin light chain kinase (MLCK) inhibitor.
Catalog No. 50-205-3748