Learn More
Enzo Life Sciences W-7 (50mg). CAS: 61714-27-0
SDP
Catalog No. 502010387
Encompass_Preferred
Binds to calmodulin, inhibiting Ca2+-calmodulin-regulated enzyme activity. It inhibits the Ca2+/calmodulin-induced activation of myosin light chain kinase in a concentration dependent manner (IC50=25 µM). Cell permeable. Alternative name: N-(6-Aminohexyl)-5-chloro-1-naphtalene-sulfonamide . HCl. Purity: ≥98% (TLC). Solubility: Soluble in water (warm), methanol, 100% ethanol (10mg/ml) (warm), DMSO (25mg/ml), dimethyl formamide or ethylene glycol. Long Term Storage: -20°C.
Catalog No. 50-201-0387 Supplier Enzo Life Sciences Supplier No. BMLCA3200050
Binds to calmodulin, inhibiting Ca2+-calmodulin-regulated enzyme activity. It inhibits the Ca2+/calmodulin-induced activation of myosin light chain kinase in a concentration dependent manner (IC50=25 µM). Cell permeable. Alternative name: N-(6-Aminohexyl)-5-chloro-1-naphtalene-sulfonamide . HCl. Purity: ≥98% (TLC). Solubility: Soluble in water (warm), methanol, 100% ethanol (10mg/ml) (warm), DMSO (25mg/ml), dimethyl formamide or ethylene glycol. Long Term Storage: -20°C.