Learn More
Medchemexpress LLC HY-100912 25mg Medchemexpress, W-7 (hydrochloride) CAS:61714-27-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10091225MG
Encompass_Preferred
Medchemexpress, HY-100912 25mg W-7 (hydrochloride) CAS:61714-27-0 W-7 hydrochloride is a selective calmodulin antagonist. W-7 hydrochloride inhibits the Ca2+-calmodulin-dependent phosphodiesterase and myosin light chain kinase with IC50 values of 28 μM and 51 µM, respectively[2]. W-7 hydrochloride induces apoptosis and has antitumor activity[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9478