Learn More
Selleck Chemical LLC W-7 hydrochloride E4636-5MG
SDP
Catalog No. 5000038952
Encompass_Preferred
W-7 hydrochloride is a potent antagonist of calmodulin (CaM) that inhibits Dengue virus (DENV) translation and replication It also competitively inhibits Trypsin-treated Ca2 /calmodulin-dependent phosphodiesterase (Ca2 -PDE) It exhibits anti-proliferative and anti-tumor effect
Catalog No. 50-000-38952 Supplier Selleck Chemical LLC Supplier No. E46365MG
W-7 hydrochloride is a potent antagonist of calmodulin (CaM) that inhibits Dengue virus (DENV) translation and replication It also competitively inhibits Trypsin-treated Ca2 /calmodulin-dependent phosphodiesterase (Ca2 -PDE) It exhibits anti-proliferative and anti-tumor effect