Learn More
Medchemexpress LLC W-7 isomer hydrochloride | 69762-85-2 | 99.7% | 377.33 | C16H22Cl2N2O2S | 100 MG
SDP
Catalog No. 5000213004
Encompass_Preferred
W-7 isomer hydrochloride is the hydrochloride salt of a W-7 isomer used as a selective calmodulin antagonist in biochemical and cell biology research. It is provided as a purified solid or as a DMSO solution for small-scale laboratory experiments and mechanistic studies of Ca2+/calmodulin-dependent processes.
  • Selective calmodulin antagonist suitable for biochemical assays and cell signaling studies.
  • High purity, 99.7%, for reproducible experimental results.
  • Available as powder and as a 10 mM solution in DMSO for ready-to-use applications.
  • Small pack sizes support pilot experiments and dose-response testing.
  • Long-term stability when stored at recommended low temperatures.
Catalog No. 50-002-13004 Supplier Medchemexpress LLC Supplier No. HY100913100MG
W-7 isomer hydrochloride is the hydrochloride salt of a W-7 isomer used as a selective calmodulin antagonist in biochemical and cell biology research. It is provided as a purified solid or as a DMSO solution for small-scale laboratory experiments and mechanistic studies of Ca2+/calmodulin-dependent processes.
  • Selective calmodulin antagonist suitable for biochemical assays and cell signaling studies.
  • High purity, 99.7%, for reproducible experimental results.
  • Available as powder and as a 10 mM solution in DMSO for ready-to-use applications.
  • Small pack sizes support pilot experiments and dose-response testing.
  • Long-term stability when stored at recommended low temperatures.