Learn More
Medchemexpress LLC W-7 (isomer hydrochloride) | 69762-85-2 | 99.7% | 377.33 | C16H22Cl2N2O2S | 50 MG
SDP
Catalog No. 5000213000
Encompass_Preferred
W-7 (isomer hydrochloride) is a hydrochloride salt provided as a solid research reagent that acts as a calmodulin antagonist. It is used in biochemical and cellular studies to probe calmodulin-dependent signaling and enzyme activity; handle and store according to the supplier SDS for safety and stability.
Catalog No. 50-002-13000 Supplier Medchemexpress LLC Supplier No. HY10091350MG
W-7 (isomer hydrochloride) is a hydrochloride salt provided as a solid research reagent that acts as a calmodulin antagonist. It is used in biochemical and cellular studies to probe calmodulin-dependent signaling and enzyme activity; handle and store according to the supplier SDS for safety and stability.