Learn More
Medchemexpress LLC W-7 isomer hydrochloride | 69762-85-2 | 99.7% | 377.33 g·mol⁻1 | C16H22Cl2N2O2S | 1 MG
SDP
Catalog No. 5000212594
Encompass_Preferred
W-7 isomer hydrochloride is the positional isomer of W-7 hydrochloride, a selective calmodulin antagonist. Provided for research use, the compound is supplied as a solid in multiple sizes and as a DMSO solution, and is intended for studies probing calcium-dependent signaling where calmodulin inhibition is required.
  • High purity (99.65%).
  • Molecular weight 377.33 g·mol⁻1.
  • Available as solid and 10 mM DMSO solution.
  • Offered in multiple pack sizes from milligram quantities to larger amounts.
  • Documents available: data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-12594 Supplier Medchemexpress LLC Supplier No. HY1009131MG
W-7 isomer hydrochloride is the positional isomer of W-7 hydrochloride, a selective calmodulin antagonist. Provided for research use, the compound is supplied as a solid in multiple sizes and as a DMSO solution, and is intended for studies probing calcium-dependent signaling where calmodulin inhibition is required.
  • High purity (99.65%).
  • Molecular weight 377.33 g·mol⁻1.
  • Available as solid and 10 mM DMSO solution.
  • Offered in multiple pack sizes from milligram quantities to larger amounts.
  • Documents available: data sheet, certificate of analysis, and safety data sheet.