Learn More
Cayman Chemical W-7hydrochlorIde 10mg
SDP
Catalog No. 502970809
Encompass_Preferred
A cell-permeable antagonist of calmodulin (Ki = 11 μM); also associates, at lower affinities, with calcium-binding domains of other proteins, including troponin C and myosin light chain kinase (Ki = 70 and 300 μM, respectively)
Catalog No. 50-297-0809 Supplier Cayman Chemical Supplier No. 1482610 MG
A cell-permeable antagonist of calmodulin (Ki = 11 μM); also associates, at lower affinities, with calcium-binding domains of other proteins, including troponin C and myosin light chain kinase (Ki = 70 and 300 μM, respectively)