Learn More
Cayman Chemical W-7 (hydrochloride), Purity: ≥98%, CAS: 61714-27-0, 50 mg
SDP
Catalog No. 502970080
Encompass_Preferred
A cell-permeable antagonist of calmodulin (Ki = 11 μM); also associates, at lower affinities, with calcium-binding domains of other proteins, including troponin C and myosin light chain kinase (Ki = 70 and 300 μM, respectively)
Catalog No. 50-297-0080 Supplier Cayman Chemical Supplier No. 1482650 MG
A cell-permeable antagonist of calmodulin (Ki = 11 μM); also associates, at lower affinities, with calcium-binding domains of other proteins, including troponin C and myosin light chain kinase (Ki = 70 and 300 μM, respectively)