Learn More
Medchemexpress LLC W-84 dibromide | 21093-51-6 | MFCD00209827 | 98.0% | 708.52 g/mol | C32H44Br2N4O4 | 5 MG
SDP
Catalog No. 5000212983
Encompass_Preferred
W-84 dibromide is a dibromide salt used as a non-competitive allosteric modulator of M2 muscarinic acetylcholine receptors for research applications. It stabilizes cholinergic antagonist-receptor complexes and is employed in studies of receptor pharmacology and as an adjunct in organophosphate protection experiments. The compound is supplied as a solid with documented purity and storage recommendations.
  • Allosteric modulator of M2 muscarinic receptors.
  • Stabilizes cholinergic antagonist-receptor complexes.
  • Used in receptor pharmacology and organophosphate protection studies.
  • Supplied as a solid with documented purity (>98%).
  • Typical storage: 4°C sealed; solvent storage guidelines available.
Catalog No. 50-002-12983 Supplier Medchemexpress LLC Supplier No. HY1009795MG
W-84 dibromide is a dibromide salt used as a non-competitive allosteric modulator of M2 muscarinic acetylcholine receptors for research applications. It stabilizes cholinergic antagonist-receptor complexes and is employed in studies of receptor pharmacology and as an adjunct in organophosphate protection experiments. The compound is supplied as a solid with documented purity and storage recommendations.
  • Allosteric modulator of M2 muscarinic receptors.
  • Stabilizes cholinergic antagonist-receptor complexes.
  • Used in receptor pharmacology and organophosphate protection studies.
  • Supplied as a solid with documented purity (>98%).
  • Typical storage: 4°C sealed; solvent storage guidelines available.