Learn More
Selleck Chemical LLC W-84 Dibromide-E0382-5MG
SDP
Catalog No. 503137789
Encompass_Preferred
W-84 dibromide (HDMPPA) is a potent allosteric modulator of M2-acetylcholine receptors which retards 3HN-methylscopolamine dissociation
Catalog No. 50-313-7789 Supplier Selleck Chemical LLC Supplier No. E03825MG
W-84 dibromide (HDMPPA) is a potent allosteric modulator of M2-acetylcholine receptors which retards 3HN-methylscopolamine dissociation