Learn More
Cayman Chemical ω-azido-C6 Ceramide, Purity: ≥95%, CAS: 2108102-18-5, 10 mg
SDP
Catalog No. 502915804
Encompass_Preferred
A clickable form of C6 ceramide; active against N. meningitidis and N. gonorrhoeae (MIC = 2 µg/ml for both); has been used to conjugate alkynylated fluorescent probes to liposomes containing ω-azido-C6 ceramide via a click reaction
Catalog No. 50-291-5804 Supplier Cayman Chemical Supplier No. 3765010 MG
A clickable form of C6 ceramide; active against N. meningitidis and N. gonorrhoeae (MIC = 2 µg/ml for both); has been used to conjugate alkynylated fluorescent probes to liposomes containing ω-azido-C6 ceramide via a click reaction