Learn More
Sigma Aldrich Fine Chemicals Biosciences Western Blot Box | Clear | 8.9 cm × 6.5 cm × 2.5 cm | 5/pack
Catalog No. NC1897240
Encompass
No matter what size your Western blot is, there′s a box that fits it perfectly. With boxes that are correctly matched to common blot sizes, you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally, all corners are formed at 90° angles and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies (for example, popular mini-gels require only about 4mL).
Catalog No. NC1897240 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7420985EA
No matter what size your Western blot is, there′s a box that fits it perfectly. With boxes that are correctly matched to common blot sizes, you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally, all corners are formed at 90° angles and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies (for example, popular mini-gels require only about 4mL).