Learn More
Cayman Chemical W-Conotoxin MVIIC trifluoroa
SDP
Catalog No. 502936402
Encompass_Preferred
An N-, P-, and Q-type calcium channel blocker; blocks N-type calcium channels on rat SCGs and P-type calcium channels on rat Purkinje neurons (Kds = 30 and ~50 nM, respectively, in the presence of 5 mM barium); blocks Q-type channels in rat CA3 neurons; completely blocks calcium uptake by rat brain synaptosomes (2.5 μM in the presence of 5 mM potassium); blocks potassium-evoked dopamine release from rat striatal slices (IC50 = ~128 nM) and potassium-evoked calcium-dependent glutamate release from rat brain synaptosomes
Catalog No. 50-293-6402 Supplier Cayman Chemical Supplier No. 24411500 UG
An N-, P-, and Q-type calcium channel blocker; blocks N-type calcium channels on rat SCGs and P-type calcium channels on rat Purkinje neurons (Kds = 30 and ~50 nM, respectively, in the presence of 5 mM barium); blocks Q-type channels in rat CA3 neurons; completely blocks calcium uptake by rat brain synaptosomes (2.5 μM in the presence of 5 mM potassium); blocks potassium-evoked dopamine release from rat striatal slices (IC50 = ~128 nM) and potassium-evoked calcium-dependent glutamate release from rat brain synaptosomes