Learn More
Bellco Glass, Inc. Water Jacketed Bioreactor Vessel, 15L
SDP
Catalog No. 501946473
Encompass_Preferred
Round bottom bioreactor vessel is manufactured of type 1 class a Borosilicate glass. Graduations are 500ml to 2500ml every 500ml. Vessel dimensions are 300mm OD x 415mm OAL. A full range of accessories are available for this vessel. 15L.. MADE IN THE USA
Catalog No. 50-194-6473 Supplier Bellco Glass, Inc. Supplier No. 790911015
Round bottom bioreactor vessel is manufactured of type 1 class a Borosilicate glass. Graduations are 500ml to 2500ml every 500ml. Vessel dimensions are 300mm OD x 415mm OAL. A full range of accessories are available for this vessel. 15L.. MADE IN THE USA