Learn More
Bellco Glass, Inc. Water Jacketed Flask Complete, 250mL
SDP
Catalog No. 501945034
Encompass_Preferred
For use circulating water bath or chiller and magnetic stir plate. Hose connection accepts 3/8″ ID silicone tubing (Bellco M7965037) Available complete with micro-carrier impeller assembly or as components. MADE IN THE USA. Specifications:Jacket Volume (mL): 175mlCap Size: 70mmSidearm Cap Size (mm): 25OD x OAH: 85x160mm
Catalog No. 50-194-5034 Supplier Bellco Glass, Inc. Supplier No. 196550250
For use circulating water bath or chiller and magnetic stir plate. Hose connection accepts 3/8″ ID silicone tubing (Bellco M7965037) Available complete with micro-carrier impeller assembly or as components. MADE IN THE USA. Specifications:Jacket Volume (mL): 175mlCap Size: 70mmSidearm Cap Size (mm): 25OD x OAH: 85x160mm