Learn More
Bellco Glass, Inc. Water Jacketed Flask Complete, 500mL
SDP
Catalog No. 501945036
Encompass_Preferred
For use circulating water bath or chiller and magnetic stir plate. Hose connection accepts 3/8″ ID silicone tubing (Bellco M7965037) Available complete with micro-carrier impeller assembly or as components. MADE IN THE USA. Specifications:Jacket Volume (mL): 260mlCap Size: 70mmSidearm Cap Size (mm): 38OD x OAH: 110x200mm
Catalog No. 50-194-5036 Supplier Bellco Glass, Inc. Supplier No. 196550500
For use circulating water bath or chiller and magnetic stir plate. Hose connection accepts 3/8″ ID silicone tubing (Bellco M7965037) Available complete with micro-carrier impeller assembly or as components. MADE IN THE USA. Specifications:Jacket Volume (mL): 260mlCap Size: 70mmSidearm Cap Size (mm): 38OD x OAH: 110x200mm