Learn More
Bellco Glass, Inc. Water Jacketed Flask Only,100mL
SDP
Catalog No. 501945033
Encompass_Preferred
Flask Only. For use circulating water bath or chiller and magnetic stir plate. Hose connection accepts 3/8″ ID silicone tubing (Bellco M7965037) Available complete with micro-carrier impeller assembly or as components. MADE IN THE USA. Specifications:Jacket Volume (mL): 75mlCap Size: 70mmSidearm Cap Size (mm): 18OD x OAH: 75x160mm
Catalog No. 50-194-5033 Supplier Bellco Glass, Inc. Supplier No. 196550101
Flask Only. For use circulating water bath or chiller and magnetic stir plate. Hose connection accepts 3/8″ ID silicone tubing (Bellco M7965037) Available complete with micro-carrier impeller assembly or as components. MADE IN THE USA. Specifications:Jacket Volume (mL): 75mlCap Size: 70mmSidearm Cap Size (mm): 18OD x OAH: 75x160mm