Learn More
Bellco Glass, Inc. Water Jacketed Flask Only,3L
SDP
Catalog No. 501945041
Encompass_Preferred
Flask only. For use circulating water bath or chiller and magnetic stir plate. Hose connection accepts 3/8″ ID silicone tubing (Bellco M7965037) Available complete with micro-carrier impeller assembly or as components. MADE IN THE USA. Specifications:Jacket Volume (mL): 1300mlCap Size: 100mmSidearm Cap Size (mm): 45OD x OAH: 200x330mm
Catalog No. 50-194-5041 Supplier Bellco Glass, Inc. Supplier No. 196553001
Flask only. For use circulating water bath or chiller and magnetic stir plate. Hose connection accepts 3/8″ ID silicone tubing (Bellco M7965037) Available complete with micro-carrier impeller assembly or as components. MADE IN THE USA. Specifications:Jacket Volume (mL): 1300mlCap Size: 100mmSidearm Cap Size (mm): 45OD x OAH: 200x330mm