Learn More
Bellco Glass, Inc. Water Jacketed Flask Only,50mL
SDP
Catalog No. 501945031
Encompass_Preferred
Flask Only. For use circulating water bath or chiller and magnetic stir plate. Hose connection accepts 3/8″ ID silicone tubing (Bellco M7965037) Available complete with micro-carrier impeller assembly or as components. MADE IN THE USA. Specifications:Jacket Volume (mL): 40mlCap Size: 51mmSidearm Cap Size (mm): 18OD x OAH: 70x120mm
Catalog No. 50-194-5031 Supplier Bellco Glass, Inc. Supplier No. 196550051
Flask Only. For use circulating water bath or chiller and magnetic stir plate. Hose connection accepts 3/8″ ID silicone tubing (Bellco M7965037) Available complete with micro-carrier impeller assembly or as components. MADE IN THE USA. Specifications:Jacket Volume (mL): 40mlCap Size: 51mmSidearm Cap Size (mm): 18OD x OAH: 70x120mm