Learn More
Bellco Glass, Inc. Water Jacketed MicroCarrier Spinner Flask Complete,6L
SDP
Catalog No. 501945042
Encompass_Preferred
For use circulating water bath or chiller and magnetic stir plate. Hose connection accepts 3/8″ ID silicone tubing (Bellco M7965037) Available complete with micro-carrier impeller assembly or as components. MADE IN THE USA. Specifications:Jacket Volume (mL): 3700mlCap Size: 100mmSidearm Cap Size (mm): 45OD x OAH: 200x445mm
Catalog No. 50-194-5042 Supplier Bellco Glass, Inc. Supplier No. 196556000
For use circulating water bath or chiller and magnetic stir plate. Hose connection accepts 3/8″ ID silicone tubing (Bellco M7965037) Available complete with micro-carrier impeller assembly or as components. MADE IN THE USA. Specifications:Jacket Volume (mL): 3700mlCap Size: 100mmSidearm Cap Size (mm): 45OD x OAH: 200x445mm