Learn More
Bellco Glass, Inc. Water Jacketed MicroCarrier Spinner Flask Complete,8L
SDP
Catalog No. 501945044
Encompass_Preferred
For use circulating water bath or chiller and magnetic stir plate. Hose connection accepts 3/8″ ID silicone tubing (Bellco M7965037) Available complete with micro-carrier impeller assembly or as components. MADE IN THE USA. Specifications:Jacket Volume (mL): 7500mlCap Size: 100mmSidearm Cap Size (mm): 45OD x OAH: 300x465mm
Catalog No. 50-194-5044 Supplier Bellco Glass, Inc. Supplier No. 196558000
For use circulating water bath or chiller and magnetic stir plate. Hose connection accepts 3/8″ ID silicone tubing (Bellco M7965037) Available complete with micro-carrier impeller assembly or as components. MADE IN THE USA. Specifications:Jacket Volume (mL): 7500mlCap Size: 100mmSidearm Cap Size (mm): 45OD x OAH: 300x465mm