Learn More
Bellco Glass, Inc. Water Jacketed Split Ring Collar Assembly ,7L
SDP
Catalog No. 501946450
Encompass_Preferred
Split flange ring assembly, 7L. Includes: A555-254 - Top gasketA555-237 - Side cushionA505-613 - Thumb nuts (6)A555-22 - Teflon washers (6)B555-11 - Split ring collar (for Water Jacketed vessel)Hardware. MADE IN THE USA
Catalog No. 50-194-6450 Supplier Bellco Glass, Inc. Supplier No. 790900044
Split flange ring assembly, 7L. Includes: A555-254 - Top gasketA555-237 - Side cushionA505-613 - Thumb nuts (6)A555-22 - Teflon washers (6)B555-11 - Split ring collar (for Water Jacketed vessel)Hardware. MADE IN THE USA