Learn More
Bellco Glass, Inc. Water Jacketed Vertical Side Arm Vessel,8L- 4 Side Arms 4 Vertical Sidearms
SDP
Catalog No. 501944776
Encompass_Preferred
Ideal for use with water baths for precise temperature control. Supplied with 100mm center cap, sidearms cap, and pouring ring. MADE IN THE USA Size: 8l Material: type 1 class a Borosilicate glass Dimensions of Water Jacketed: 300mm od x 440 oah Dimension of inner flask: 238mm OD x 420mm OAH. Dimensions: width including two hose barbs 396mm. 2 - #4 hose connectors. 100mm screw thread H.W. neck. 4 - 45mm screw thread vertical side arms. Graduations:1500mL to 8000mL every 500mL
Catalog No. 50-194-4776 Supplier Bellco Glass, Inc. Supplier No. 196418040
Ideal for use with water baths for precise temperature control. Supplied with 100mm center cap, sidearms cap, and pouring ring. MADE IN THE USA Size: 8l Material: type 1 class a Borosilicate glass Dimensions of Water Jacketed: 300mm od x 440 oah Dimension of inner flask: 238mm OD x 420mm OAH. Dimensions: width including two hose barbs 396mm. 2 - #4 hose connectors. 100mm screw thread H.W. neck. 4 - 45mm screw thread vertical side arms. Graduations:1500mL to 8000mL every 500mL