Learn More
Cayman Chemical W-MurIcholIc AcId 10mg
SDP
Catalog No. 502983040
Encompass_Preferred
A murine-specific secondary bile acid; formed via enzymatic conversion of β-MCA by a variety of intestinal microorganisms in rat and mouse gut
Catalog No. 50-298-3040 Supplier Cayman Chemical Supplier No. 2029210 MG
A murine-specific secondary bile acid; formed via enzymatic conversion of β-MCA by a variety of intestinal microorganisms in rat and mouse gut