Learn More
Cayman Chemical W-MurIcholIc AcId 1mg
SDP
Catalog No. 502992658
Encompass_Preferred
A murine-specific secondary bile acid; formed via enzymatic conversion of β-MCA by a variety of intestinal microorganisms in rat and mouse gut
This item is not returnable. View return policy
Catalog No. 50-299-2658 Supplier Cayman Chemical Supplier No. 202921 MG
A murine-specific secondary bile acid; formed via enzymatic conversion of β-MCA by a variety of intestinal microorganisms in rat and mouse gut