Learn More
Cayman Chemical ω-Muricholic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502941969
Encompass_Preferred
An internal standard for the quantification of ω-MCA by GC- or LC-MS
This item is not returnable. View return policy
Catalog No. 50-294-1969 Supplier Cayman Chemical Supplier No. 270371 MG
An internal standard for the quantification of ω-MCA by GC- or LC-MS