Learn More
Cayman Chemical ω-Muricholic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), 5 mg
SDP
Catalog No. 502937924
Encompass_Preferred
An internal standard for the quantification of ω-MCA by GC- or LC-MS
Catalog No. 50-293-7924 Supplier Cayman Chemical Supplier No. 270375 MG
An internal standard for the quantification of ω-MCA by GC- or LC-MS