Learn More
W.S. TYLER™ RO-TAP™ Wet Test Kit
Catalog No. 049096 Shop All W. S. Tyler Products
Change view
Click to view available options
For Use With (Equipment):
Ro-Tap II sieve shaker
Catalog No. For Use With (Equipment)
04-909-6 Ro-Tap II sieve shaker
1 options
Catalog No. 04-909-6 Supplier W.S. TYLER™ Supplier No. LA11112
Add to Cart
Add to Cart
Includes
flowmeter assembly and stand, 90° angle connector, cover, 3-prong clamp, cork plug, sieve support, polyethylene tee, two 12 ft. (3.7m) lengths of thin-walled amber tubing, cover with two stainless-steel tubes, and 8 in. diameter (20.3cm) bottom pan with stainless-steel outlet tube.
For use with the RO-TAP II Sieve Shaker
Specifications
Product Type Wet Test Kit
Weight (English) 27 lb.
Weight (Metric) 12.24 kg
For Use With (Equipment) Ro-Tap II sieve shaker
Includes Flowmeter assembly and stand, 90 degree connector, cover, 3-prong clamp, cork plug, sieve support, polyethylene “tee,” two 12 ft. (3.7m) lengths of thin-walled amber tubing, cover with two Stainless-steel tubes and 8 in. diameter (20.3cm) bottom pan with