Learn More
Grainger W Strap Flat-Hook Orange
Catalog No. 502765600
Encompass_Preferred
The ANCRA Tie Down Strap Ratchet is a heavy duty ratchet tie down strap made of polyester webbing with a working load limit of 1,823 lbs. It features a ratchet mechanism for easy tightening and release and measures 15 ft in length and 1 in width. The strap has a flat hook on one end and a ratchet mechanism with a J-hook on the other end for attaching to anchor points. It can be used for securing cargo, vehicles, trailers, and equipment during transport and meets DOT regulations. The polyester webbing is resistant to rot, mildew, water, grease, and UV rays. This heavy duty tie down strap provides a safe and secure method for tie down and transport applications.
Catalog No. 50-276-5600 Supplier Grainger Supplier No. 16V909
The ANCRA Tie Down Strap Ratchet is a heavy duty ratchet tie down strap made of polyester webbing with a working load limit of 1,823 lbs. It features a ratchet mechanism for easy tightening and release and measures 15 ft in length and 1 in width. The strap has a flat hook on one end and a ratchet mechanism with a J-hook on the other end for attaching to anchor points. It can be used for securing cargo, vehicles, trailers, and equipment during transport and meets DOT regulations. The polyester webbing is resistant to rot, mildew, water, grease, and UV rays. This heavy duty tie down strap provides a safe and secure method for tie down and transport applications.