Learn More
Vee Gee Wide-Tip Volumetric Pipet, 25 mL
Supplier:  Vee Gee 2040253MM
Encompass_Preferred
SIBATA reusable glass Volumetric, Wide-Tip Opening, Pipets deliver. They are ideal for water treatment facilities, winemakers, tissue culture work and essential oil testing.• Meet ASTM E-969 Class B Specifications for tolerance : 25.0 mL • Tolerance: ± 0.03 mL • Approx.: 475 mm L x 3.0 mm Tip I.D.
Catalog No. NC2386467