Learn More
Alkali Scientific Alkali Scientific™ W1005-1000 M1 Grade Calibration Weight, 1000g
SDP
Catalog No. 503029562
Encompass_Preferred
Alkali Scientific™ W1005-1000 M1 Grade Calibration Weight, 1000g from economy to expensive analytical models, require routine calibration and accuracy confirmation. All individual weights are supplied in protective, screw capped plastic containers.  We offer both F1 and M1 class individual weights. Alkali Scientific™ W1005-1000 M1 Grade Calibration Weight, 1000g is supplied with an individual Certificate of Verification confirming that the weight(s) meet OIML regulations and tolerances for the specific weight class.
Catalog No. 50-302-9562 Supplier Alkali Scientific Supplier No. RS77191000
Alkali Scientific™ W1005-1000 M1 Grade Calibration Weight, 1000g from economy to expensive analytical models, require routine calibration and accuracy confirmation. All individual weights are supplied in protective, screw capped plastic containers.  We offer both F1 and M1 class individual weights. Alkali Scientific™ W1005-1000 M1 Grade Calibration Weight, 1000g is supplied with an individual Certificate of Verification confirming that the weight(s) meet OIML regulations and tolerances for the specific weight class.