Learn More
Alkali Scientific Alkali Scientific™ W1101-6-100 Class E2, Calibration set for Analytical Balances, 6 pcs: 1x100mg, 1x10g, 2x20g, 1x50g, 1x100g
SDP
Catalog No. 503029620
Encompass_Preferred
Our Alkali Scientific™ W1101-6-100 Class E2, Calibration set for Analytical Balances, 6 pcs: 1x100mg, 1x10g, 2x20g, 1x50g, 1x100g  is ideal for calibration of analytical balances with a 0.0001g readability. Precision manufactured from high purity, 316 stainless steel (7.96kg/dm³), these weights are corrosion resistant with magnetic susceptibility <0.005. The set includes 6 weights, 1x100mg, 1x10g, 2x20g, 1x50g, 1x100g, for checking repeatability, cornerload, linearity and span. The weights are supplied in a protective case with tweezers, a cleaning cloth and a certificate of verification
Catalog No. 50-302-9620 Supplier Alkali Scientific Supplier No. W11016100
Our Alkali Scientific™ W1101-6-100 Class E2, Calibration set for Analytical Balances, 6 pcs: 1x100mg, 1x10g, 2x20g, 1x50g, 1x100g  is ideal for calibration of analytical balances with a 0.0001g readability. Precision manufactured from high purity, 316 stainless steel (7.96kg/dm³), these weights are corrosion resistant with magnetic susceptibility <0.005. The set includes 6 weights, 1x100mg, 1x10g, 2x20g, 1x50g, 1x100g, for checking repeatability, cornerload, linearity and span. The weights are supplied in a protective case with tweezers, a cleaning cloth and a certificate of verification