Learn More
Andwin Scientific W1126 WEIGH BOATS 100/PK
SDP
Catalog No. NC9877633
Encompass_Preferred
The aluminum weighing boats have a rectangular shape and flat bottom. The boats have a dimension of 4 mm W 12 mm L 4 mm D. This product also features tweezer-grip tab that allows easy handling. This is recommended for use for electrobalance weighing and combustion analysis. It is sold in a pack of 100.
Catalog No. NC9877633 Supplier Andwin Scientific Supplier No. W1126100EA
The aluminum weighing boats have a rectangular shape and flat bottom. The boats have a dimension of 4 mm W 12 mm L 4 mm D. This product also features tweezer-grip tab that allows easy handling. This is recommended for use for electrobalance weighing and combustion analysis. It is sold in a pack of 100.