Learn More
Cayman Chemical W140 (trifluoroacetate salt), Purity: ≥95%, CAS: 909725-64-0, 1 mg
SDP
Catalog No. 502890363
Encompass_Preferred
An inactive enantiomer of W146; binds weakly to S1P1 (Ki = 4.6 µM for the human receptor); has been used as a negative control in the study of S1P1 receptor signaling
Catalog No. 50-289-0363 Supplier Cayman Chemical Supplier No. 100091101 MG
An inactive enantiomer of W146; binds weakly to S1P1 (Ki = 4.6 µM for the human receptor); has been used as a negative control in the study of S1P1 receptor signaling