Learn More
Cayman Chemical W140 trifluoroacetate salt
Supplier: Cayman Chemical 10009110500 UG
An inactive enantiomer of W146; binds weakly to S1P1 (Ki = 4.6 µM for the human receptor); has been used as a negative control in the study of S1P1 receptor signaling