Learn More
Medchemexpress LLC W146 trifluoroacetate | 909725-62-8 | 98.2% | 456.39 g/mol | C18H28F3N2O6P | 25 MG
SDP
Catalog No. 5000212835
Encompass_Preferred
W146 trifluoroacetate is a research-grade selective antagonist of sphingosine-1-phosphate receptor 1 (S1PR1). Supplied as the trifluoroacetate salt in a white to off-white solid form, it has an EC50 of 398 nM, a molecular weight of 456.39 g/mol, and is typically supplied at high purity for in vitro pharmacology and receptor signaling studies. Not for human or clinical use.
  • Selective S1PR1 antagonist with EC50 398 nM
  • Provided as trifluoroacetate salt for stability
  • High purity suitable for biochemical assays
  • White to off-white solid for easy handling
  • Intended for in vitro pharmacology and receptor signaling studies
Catalog No. 50-002-12835 Supplier Medchemexpress LLC Supplier No. HY101395A25MG
W146 trifluoroacetate is a research-grade selective antagonist of sphingosine-1-phosphate receptor 1 (S1PR1). Supplied as the trifluoroacetate salt in a white to off-white solid form, it has an EC50 of 398 nM, a molecular weight of 456.39 g/mol, and is typically supplied at high purity for in vitro pharmacology and receptor signaling studies. Not for human or clinical use.
  • Selective S1PR1 antagonist with EC50 398 nM
  • Provided as trifluoroacetate salt for stability
  • High purity suitable for biochemical assays
  • White to off-white solid for easy handling
  • Intended for in vitro pharmacology and receptor signaling studies