Learn More
Cayman Chemical W146 trifluoroacetate salt
SDP
Supplier:  Cayman Chemical 1000910910 MG
Encompass_Preferred
An S1P1 receptor antagonist (Ki = 77 nM); selective for S1P1 over S1P2, S1P3, and S1P5 (Kis = >10 µM); prevents ligand-induced receptor internalization in CHO cells and inhibits ERK and Akt phosphorylation at 10 µM; increases AMD3100-induced KSL-HSPC entrance into mouse blood circulation from the bone marrow at 5 mg/kg in combination with AMD3100; disrupts the endothelial barrier and induces PMN neutrophil infiltration into the lungs and skin in an ovalbumin-induced mouse model of immune cell-mediated vascular injury at 10 UG intranasally or intradermally
Catalog No. 50-289-1268