Learn More
Alkali Scientific W3100-210 Analytical Balance 2
SDP
Catalog No. 503029501
Encompass_Preferred
Alkali ScientificW3100-210 Analytical Balance 210 grams readability 0 0001grams 115V incorporate electromagnetic sensor technology and precision mechanical components to offer superior weight determinations to a fraction of a milligram Two capacity ranges are available in the Analytical Balance line to weigh samples up to 120g or 210g Readability for both ranges is 0 0001g (0 1mg) with accuracy and linearity specifications that match up with the top brands in the industry The W3100 balances tare and stabilize quickly to provide immediate results Readablity to 0 0001g (0 1 mg) Available with Quick-Calinternal calibration feature RS232 port for data output to a computer or printer Multiple mode options (standard weighing count mode percent) Both ranges offer an optional Quick-CalTM internal calibration feature that allows quick and convenient calibration without the need for a calibration standard weight The glass draft shield enclosing the weighing chamber has sliding doors on
Catalog No. 50-302-9501 Supplier Alkali Scientific Supplier No. W3100210
Alkali ScientificW3100-210 Analytical Balance 210 grams readability 0 0001grams 115V incorporate electromagnetic sensor technology and precision mechanical components to offer superior weight determinations to a fraction of a milligram Two capacity ranges are available in the Analytical Balance line to weigh samples up to 120g or 210g Readability for both ranges is 0 0001g (0 1mg) with accuracy and linearity specifications that match up with the top brands in the industry The W3100 balances tare and stabilize quickly to provide immediate results Readablity to 0 0001g (0 1 mg) Available with Quick-Calinternal calibration feature RS232 port for data output to a computer or printer Multiple mode options (standard weighing count mode percent) Both ranges offer an optional Quick-CalTM internal calibration feature that allows quick and convenient calibration without the need for a calibration standard weight The glass draft shield enclosing the weighing chamber has sliding doors on