Learn More
Alkali Scientific W3200-5K Precision Balance 500
SDP
Catalog No. 503029578
Encompass_Preferred
Alkali ScientificW3200-5K Precision Balance quickly and accurately display sample measurements on a large backlit LCD screen Utilizing highly precise gold plated ceramic capacitance sensors these balances tare and stabilize quickly and provide reliable weight measurements within 0 001 grams Six models are available in the Precision Balance range covering a wide range of maximum capacity requirements The smaller capacity models (120g 320g and 500g) include a glass draft shield with 3 sliding doors for easy access the weighing chamber These models offer readability to 001g (1mg) The larger capacity models (1200g 3200g and 5000g) feature a more traditional top-loading design come with larger weigh pans and have readability to 0 01g All models feature a control panel with a large bright back-lit LCD screen that is easy to use and easy to see The advanced software includes weighing modes for standard weighing piece counting percent deviation and also check weighing for component insp
Catalog No. 50-302-9578 Supplier Alkali Scientific Supplier No. W32005K
Alkali ScientificW3200-5K Precision Balance quickly and accurately display sample measurements on a large backlit LCD screen Utilizing highly precise gold plated ceramic capacitance sensors these balances tare and stabilize quickly and provide reliable weight measurements within 0 001 grams Six models are available in the Precision Balance range covering a wide range of maximum capacity requirements The smaller capacity models (120g 320g and 500g) include a glass draft shield with 3 sliding doors for easy access the weighing chamber These models offer readability to 001g (1mg) The larger capacity models (1200g 3200g and 5000g) feature a more traditional top-loading design come with larger weigh pans and have readability to 0 01g All models feature a control panel with a large bright back-lit LCD screen that is easy to use and easy to see The advanced software includes weighing modes for standard weighing piece counting percent deviation and also check weighing for component insp