Learn More
Alkali Scientific W3300-300 Compact Balance 300
SDP
Catalog No. 503029492
Encompass_Preferred
Alkali ScientificW3300-300 compact balances has been designed to meet the basic needs of academic and research labs requiring weight determinations to increments of 0 01g to 0 1 g The compact line includes 6 models covering a wide range of maximum weight capacities The lower capacity units (120g 300g and 500g) are readable to 0 01 g and come with a draft shield ring to prevent air currents from affecting measurements Alkali ScientificW3300-300 compact line offer large brightly lit LCD panels and are easy to operate using the intuitive and splash resistant control panel Different modes allow weight to be displayed in various units grams ounces carats and simple programs can be used for piece counting and percent deviation weighing Leveling feet are located toward the front of the balance for easy access and a leveling bubble in the control panel is easy to see for quick and simple leveling on the lab bench The balances can be calibrated using a calibration weight and a few key str
Catalog No. 50-302-9492 Supplier Alkali Scientific Supplier No. W3300C300
Alkali ScientificW3300-300 compact balances has been designed to meet the basic needs of academic and research labs requiring weight determinations to increments of 0 01g to 0 1 g The compact line includes 6 models covering a wide range of maximum weight capacities The lower capacity units (120g 300g and 500g) are readable to 0 01 g and come with a draft shield ring to prevent air currents from affecting measurements Alkali ScientificW3300-300 compact line offer large brightly lit LCD panels and are easy to operate using the intuitive and splash resistant control panel Different modes allow weight to be displayed in various units grams ounces carats and simple programs can be used for piece counting and percent deviation weighing Leveling feet are located toward the front of the balance for easy access and a leveling bubble in the control panel is easy to see for quick and simple leveling on the lab bench The balances can be calibrated using a calibration weight and a few key str