Learn More
Cold Chain Technologies W50-Uk/USA 48x40x48in or 120x100x122CM high
Catalog No. 502006799
Encompass_Preferred
Thermal Pallet Cover, Temperature Control, Breathable, Solar Radiation Reflective, Water and Wind Resistant, Tyvek by Dupont and Cold Chain Technologies, One Piece pallet cover with elastic bottom with metalized internal layer and fleece liner
Catalog No. 50-200-6799 Supplier Cold Chain Technologies Supplier No. D15519198
Thermal Pallet Cover, Temperature Control, Breathable, Solar Radiation Reflective, Water and Wind Resistant, Tyvek by Dupont and Cold Chain Technologies, One Piece pallet cover with elastic bottom with metalized internal layer and fleece liner