Learn More
Cold Chain Technologies W50-Uk/USA 52x42x48in or 130x107x122CM high
Catalog No. 502006801
Encompass_Preferred
Thermal Pallet Cover, Temperature Control, Breathable, Solar Radiation Reflective, Water and Wind Resistant, Tyvek by Dupont and Cold Chain Technologies, One Piece pallet cover with elastic bottom with metalized internal layer and fleece liner
Catalog No. 50-200-6801 Supplier Cold Chain Technologies Supplier No. D15526931
Thermal Pallet Cover, Temperature Control, Breathable, Solar Radiation Reflective, Water and Wind Resistant, Tyvek by Dupont and Cold Chain Technologies, One Piece pallet cover with elastic bottom with metalized internal layer and fleece liner