Learn More
Medchemexpress LLC WAAG-3R TFA | 99.8% | 1644.74 | 1 MG
SDP
Catalog No. 5000396317
Encompass_Preferred
WAAG-3R TFA is a biologically active peptide. Aggrecanases, which are part of the ADAMTS (A disintegrin and metalloprotease with thrombospondin motif) family of proteases, cleave aggrecan, a main structural component of cartilage. Aggrecanase-1 (ADAMTS-4) is a significant aggrecanase in human osteoarthritic cartilage. This FRET peptide was utilized in an ADAMTS-4 (Aggrecanase-1) and ADAMTS-5 (Aggrecanase-2) assay with Ex/Em = 340/420 nm.
  • Biologically active peptide.
  • Utilized in an ADAMTS-4 (Aggrecanase-1) and ADAMTS-5 (Aggrecanase-2) assay.
  • FRET peptide with Ex/Em = 340/420 nm.
Catalog No. 50-003-96317 Supplier Medchemexpress LLC Supplier No. HYP5419A1MG
WAAG-3R TFA is a biologically active peptide. Aggrecanases, which are part of the ADAMTS (A disintegrin and metalloprotease with thrombospondin motif) family of proteases, cleave aggrecan, a main structural component of cartilage. Aggrecanase-1 (ADAMTS-4) is a significant aggrecanase in human osteoarthritic cartilage. This FRET peptide was utilized in an ADAMTS-4 (Aggrecanase-1) and ADAMTS-5 (Aggrecanase-2) assay with Ex/Em = 340/420 nm.
  • Biologically active peptide.
  • Utilized in an ADAMTS-4 (Aggrecanase-1) and ADAMTS-5 (Aggrecanase-2) assay.
  • FRET peptide with Ex/Em = 340/420 nm.