Learn More
Ted Pella Inc Wafer Carrier Tray for 6"/150mm Single Wafer, Natural Polypropylene. ø160 x 11mm (Pack 10)
SDP
Catalog No. NC1125553
Encompass
These high quality wafer carrier trays are specifically designed for storage, transport and handling of semiconductor wafers, glass wafers or other delicate flat objects. Made from either natural or conductive black polypropylene material and available in sizes from 1" to 6" with maximum grip, minimum contact and minimum pressure. The wafer or disc carriers have a conical base to support the wafers at the edges only. The wafers are held in place and secured by a spider spring and locking cap.
Catalog No. NC1125553 Supplier Ted Pella Inc Supplier No. 139560
These high quality wafer carrier trays are specifically designed for storage, transport and handling of semiconductor wafers, glass wafers or other delicate flat objects. Made from either natural or conductive black polypropylene material and available in sizes from 1" to 6" with maximum grip, minimum contact and minimum pressure. The wafer or disc carriers have a conical base to support the wafers at the edges only. The wafers are held in place and secured by a spider spring and locking cap.