Learn More
Electron Microscopy Sciences Wafer Cleaving Kit with Mat
SDP
Catalog No. 501928733
Encompass_Preferred
The complete tool set to scribe and cleave cleanly. This is a critical step to getting the best cross-sections. The kit includes two diamond scribes for marking and scribing (straight tip and 30 degree tip), a pen-style diamond scribe, tweezers with black soft fiber fine tip (length 6.25 in), wafer cleaning pliers, a clear plastic ruler with metric and US units, and tungsten cleaving wire. This kit is suitable for a wide variety of substrates and wafers (Si, GaAs, glass). Includes 12x18 in wafer mat.
Catalog No. 50-192-8733 Supplier Electron Microscopy Sciences Supplier No. 7642M
The complete tool set to scribe and cleave cleanly. This is a critical step to getting the best cross-sections. The kit includes two diamond scribes for marking and scribing (straight tip and 30 degree tip), a pen-style diamond scribe, tweezers with black soft fiber fine tip (length 6.25 in), wafer cleaning pliers, a clear plastic ruler with metric and US units, and tungsten cleaving wire. This kit is suitable for a wide variety of substrates and wafers (Si, GaAs, glass). Includes 12x18 in wafer mat.