Learn More
Electron Microscopy Sciences CleanBreak Pliers | 6in
SDP
Catalog No. NC1545648
Encompass_Preferred
CleanBreak 6in wafer cleaving pliers are a simple and clean way to cleave (post scribe) wafers, wafer strips and smaller pieces. These pliers have a 3/4in jawopening and comes with one set of replaceable jaws to maintain the best cleaving performance.
Catalog No. NC1545648 Supplier Electron Microscopy Sciences Supplier No. 7646
CleanBreak 6in wafer cleaving pliers are a simple and clean way to cleave (post scribe) wafers, wafer strips and smaller pieces. These pliers have a 3/4in jawopening and comes with one set of replaceable jaws to maintain the best cleaving performance.