Learn More
Electron Microscopy Sciences Wafer Cleaving Station
SDP
Catalog No. 501902666
Encompass_Preferred
The Cleaving Station includes two diamond scribes for marking and scribing, one pen style diamond scribe and pliers for cleaving, everything in the Cleaving Kit. In addition, the Lattice Scriber, customized with an 8 point diamond tip that scribes well at any angle and small sample cleaving pliers are included so that a clean cleave is always possible on samples from a few mm's to 300 mm. The standard configuration includes one each:
Pen style, straight tip, and 30 degree tip Diamond Scribes Tweezers with black soft fiber fine tip
CleanBreak waferand small sample cleaving pliers
Small self-healing ruler mat and wafer piece ruler mat
Clear plastic ruler with metric and US units
Tungsten cleaving wire
LatticeScriber diamond scriber customized with and 8 point diamond tip
Catalog No. 50-190-2666 Supplier Electron Microscopy Sciences Supplier No. 7648
The Cleaving Station includes two diamond scribes for marking and scribing, one pen style diamond scribe and pliers for cleaving, everything in the Cleaving Kit. In addition, the Lattice Scriber, customized with an 8 point diamond tip that scribes well at any angle and small sample cleaving pliers are included so that a clean cleave is always possible on samples from a few mm's to 300 mm. The standard configuration includes one each:
Pen style, straight tip, and 30 degree tip Diamond Scribes Tweezers with black soft fiber fine tip
CleanBreak waferand small sample cleaving pliers
Small self-healing ruler mat and wafer piece ruler mat
Clear plastic ruler with metric and US units
Tungsten cleaving wire
LatticeScriber diamond scriber customized with and 8 point diamond tip