Learn More
Electron Microscopy Sciences Transparent Wafer Container Single, 3", Polypropylene Material, 84 x 11 mm
SDP
Catalog No. 50364966
Encompass_Preferred
The carriers are used for storage and or shipping wafers. They prevent all types of damage and contamination to the wafer during shipping and even storage. Each of the containers are packed in 1000 class clean room and bag. The set includes the Container , the cover and the spring. Boxes are transparent and made from Anti-Electrostatic Polycarbonate.
Use: 3" (75 mm)
Material: Polypropylene
Dimensions: 84 x 11 mm
Catalog No. 50-364-966 Supplier Electron Microscopy Sciences Supplier No. 649173
The carriers are used for storage and or shipping wafers. They prevent all types of damage and contamination to the wafer during shipping and even storage. Each of the containers are packed in 1000 class clean room and bag. The set includes the Container , the cover and the spring. Boxes are transparent and made from Anti-Electrostatic Polycarbonate.
Use: 3" (75 mm)
Material: Polypropylene
Dimensions: 84 x 11 mm